Assay Details
| Gene Symbol: Cabp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138948.3 | NP_620398.1 | 4 - 5 | 564 | 118 | |
| GenBank mRNA | AF419324.1 | - | 4 - 5 | 565 | 118 | |
| BC104356.1 | - | 4 - 5 | 563 | 40049623 | 118 | |
| BC104357.1 | - | 4 - 5 | 563 | 40049625 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559369 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559364 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559370 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559363 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559366 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559365 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033225 | NM_138948.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033225 | AF419324.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033225 | BC104356.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033225 | BC104357.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677561 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179661 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39597 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134085 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033225 | NM_138948.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033225 | AF419324.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033225 | BC104356.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033225 | BC104357.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27559369 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134085 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02252439 | - | - |
| Whole Mouse Genome G4122A | A_51_P417825 | NM_138948.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417825 | AF419324.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417825 | BC104356.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417825 | BC104357.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100009 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129901 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134085 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09557834 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09650325 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09209091 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09650324 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559363 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559365 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559364 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559370 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559366 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27559369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27559345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09619653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08989860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09198415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09619649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09538317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09206701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09206693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09688941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09206690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09597803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09557832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09147241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09597788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09557834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09619630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09619650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09650325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09619647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09147256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09538318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27559305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10383896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383892 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436141_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425963_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10383891 | - | - |
| Mouse Genome 430 2.0 Array | 1425963_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425963_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192650_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192650_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100020583.mm.1 | NM_138948.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020583.mm.1 | NM_138948.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020583.mm.1 | TC1100002120.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425963_PM_at | - | - |
| Mouse Expression Set 430 | 1425963_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425963_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425963_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425963_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425963_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 531351 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3840403 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3840403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003840403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2599292 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2599292 | - | - |
| mouse-6 v1.1 (Illumina) | scl40579.4.1_15-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3840403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40579.4.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2599292 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2599292 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40579.4.1_15-S | - | - |

Cabp7
Genomic Map