Assay Details
| Gene Symbol: Tcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130458.1 | NP_001123930.1 | 4 - 5 | 628 | 95 | |
| NM_001130459.1 | NP_001123931.1 | 4 - 5 | 597 | 95 | ||
| NM_015749.3 | NP_056564.1 | 4 - 5 | 631 | 95 | ||
| GenBank mRNA | AF090686.1 | - | 3 - 4 | 558 | 95 | |
| AK133707.1 | - | 4 - 5 | 593 | 95 | ||
| AK143761.1 | - | 4 - 5 | 599 | 95 | ||
| AK147176.1 | - | 4 - 5 | 602 | 95 | ||
| AK161618.1 | - | 4 - 5 | 598 | 95 | ||
| AK170401.1 | - | 4 - 5 | 617 | 95 | ||
| BC003720.1 | - | 3 - 4 | 504 | 3601531 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcn2
Genomic Map