Assay Details
| Gene Symbol: Mtfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026443.4 | NP_080719.2 | 1 - 2 | 217 | 81 | |
| GenBank mRNA | AK006148.1 | - | 1 - 2 | 138 | 81 | |
| AK010971.1 | - | 1 - 2 | 174 | 81 | ||
| AK012058.1 | - | 1 - 2 | 184 | 81 | ||
| AK078381.1 | - | 1 - 2 | 218 | 81 | ||
| AK167990.1 | - | 1 - 2 | 84 | 81 | ||
| BC019205.1 | - | 1 - 2 | 227 | 81 | ||
| BC058737.1 | - | 1 - 2 | 167 | 5400435 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtfp1
Genomic Map