Assay Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029291.1 | NP_083567.1 | 16 - 17 | 1824 | 80 | |
| GenBank mRNA | AK051387.1 | - | 15 - 16 | 1753 | 80 | |
| AK148105.1 | - | 16 - 17 | 1856 | 80 | ||
| BC013537.1 | - | 16 - 17 | 1824 | 4240784 | 80 | |
| BC024840.1 | - | 9 - 10 | 975 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ascc2
Genomic Map