Assay Details
| Gene Symbol: Cdc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243769.1 | NP_001230698.1 | 3 - 4 | 339 | 74 | |
| NM_009861.3 | NP_033991.1 | 3 - 4 | 339 | 74 | ||
| GenBank mRNA | AK003098.1 | - | 3 - 4 | 260 | 74 | |
| AK051543.1 | - | 3 - 4 | 260 | 74 | ||
| AK075567.1 | - | 3 - 4 | 258 | 74 | ||
| AK144216.1 | - | 3 - 4 | 262 | 74 | ||
| AK145659.1 | - | 2 - 3 | 103 | 74 | ||
| AK151087.1 | - | 3 - 4 | 284 | 74 | ||
| AK151726.1 | - | 3 - 4 | 261 | 74 | ||
| AK153564.1 | - | 3 - 4 | 283 | 74 | ||
| AK154870.1 | - | 3 - 4 | 258 | 74 | ||
| AK159470.1 | - | 3 - 4 | 301 | 74 | ||
| AK166281.1 | - | 3 - 4 | 258 | 74 | ||
| AK167195.1 | - | 3 - 4 | 247 | 74 | ||
| AK167400.1 | - | 3 - 4 | 252 | 74 | ||
| AK167609.1 | - | 3 - 4 | 257 | 74 | ||
| AK168013.1 | - | 2 - 3 | 252 | 74 | ||
| AK168076.1 | - | 3 - 4 | 252 | 74 | ||
| AK168089.1 | - | 2 - 3 | 252 | 74 | ||
| AK168276.1 | - | 3 - 4 | 252 | 74 | ||
| AK168503.1 | - | 3 - 4 | 273 | 74 | ||
| AK168758.1 | - | 3 - 4 | 292 | 74 | ||
| AK168820.1 | - | 3 - 4 | 265 | 74 | ||
| AK169122.1 | - | 3 - 4 | 273 | 74 | ||
| AK169232.1 | - | 3 - 4 | 252 | 74 | ||
| AK169805.1 | - | 3 - 4 | 277 | 74 | ||
| CD551112.1 | - | 3 - 4 | 326 | 74 | ||
| L11318.1 | - | 2 - 3 | 175 | 74 | ||
| L78075.1 | - | 2 - 3 | 179 | 74 | ||
| U37720.1 | - | 3 - 4 | 247 | 74 | ||
More Information
Set Membership: |
|

Cdc42
Genomic Map