Assay Details
| Gene Symbol: Emp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146346.1 | NP_001139818.1 | 3 - 4 | 437 | 71 | |
| NM_010129.2 | NP_034259.2 | 3 - 4 | 408 | 71 | ||
| GenBank mRNA | AF011750.1 | - | 3 - 4 | 269 | 71 | |
| AK146939.1 | - | 3 - 4 | 440 | 71 | ||
| BC001999.1 | - | 3 - 4 | 253 | 3485948 | 71 | |
| CK022835.1 | - | 3 - 4 | 407 | 71 | ||
| U87948.1 | - | 3 - 4 | 386 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Emp3
Genomic Map