Assay Details
| Gene Symbol: Sgsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162965.1 | NP_001156437.1 | 13 - 14 | 1525 | 80 | |
| NM_001254731.1 | NP_001241660.1 | 13 - 14 | 1525 | 80 | ||
| NM_172718.3 | NP_766306.2 | 13 - 14 | 1525 | 80 | ||
| NR_027934.1 | 4 - 5 | 712 | 80 | |||
| NR_027936.1 | 4 - 5 | 712 | 80 | |||
| GenBank mRNA | AK010756.1 | - | 4 - 5 | 698 | 80 | |
| AK044716.1 | - | 13 - 14 | 1479 | 80 | ||
| AK051018.1 | - | 4 - 5 | 713 | 80 | ||
| AK053555.1 | - | 13 - 14 | 1459 | 80 | ||
| AK167632.1 | - | 4 - 5 | 676 | 80 | ||
| AK170021.1 | - | 13 - 14 | 1517 | 80 | ||
| BC138049.1 | - | 13 - 14 | 1451 | 8861068 | 80 | |
| BC138050.1 | - | 13 - 14 | 1451 | 8861069 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm1
Genomic Map