Assay Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023799.3 | NP_076288.1 | 10 - 11 | 2148 | 151 | |
| GenBank mRNA | AF132214.1 | - | 10 - 11 | 1993 | 151 | |
| AK129188.1 | - | 10 - 11 | 2377 | 151 | ||
| BC054821.1 | - | 10 - 11 | 2148 | 6515250 | 151 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgea5
Genomic Map