Assay Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023799.3 | NP_076288.1 | 14 - 15 | 2616 | 69 | |
| GenBank mRNA | AF132214.1 | - | 14 - 15 | 2461 | 69 | |
| AK014781.1 | - | 2 - 3 | 242 | 69 | ||
| AK077613.1 | - | 2 - 3 | 1138 | 69 | ||
| AK129188.1 | - | 14 - 15 | 2845 | 69 | ||
| BC054821.1 | - | 14 - 15 | 2616 | 6515250 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgea5
Genomic Map