Assay Details
| Gene Symbol: Ldb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113408.1 | NP_001106879.1 | 3 - 4 | 533 | 109 | |
| NM_010697.1 | NP_034827.1 | 3 - 4 | 718 | 109 | ||
| GenBank mRNA | AB250383.1 | - | 2 - 3 | 72 | 109 | |
| AF030333.1 | - | 3 - 4 | 236 | 109 | ||
| AK193238.1 | - | 2 - 3 | 75 | 109 | ||
| BC013624.1 | - | 3 - 4 | 358 | 3708169 | 109 | |
| CK790183.1 | - | 3 - 4 | 538 | 109 | ||
| U69270.1 | - | 3 - 4 | 718 | 109 | ||
| U70375.1 | - | 2 - 3 | 68 | 109 | ||
| U89488.1 | - | 3 - 4 | 316 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldb1
Genomic Map