Assay Details
| Gene Symbol: Ldb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113408.1 | NP_001106879.1 | 9 - 10 | 1216 | 73 | |
| NM_010697.1 | NP_034827.1 | 9 - 10 | 1401 | 73 | ||
| GenBank mRNA | AB221569.1 | - | 8 - 9 | 758 | 73 | |
| AB250383.1 | - | 8 - 9 | 755 | 73 | ||
| AF030333.1 | - | 9 - 10 | 919 | 73 | ||
| AK048463.1 | - | 4 - 5 | 441 | 73 | ||
| BC013624.1 | - | 9 - 10 | 1041 | 3708169 | 73 | |
| U69270.1 | - | 9 - 10 | 1401 | 73 | ||
| U70375.1 | - | 8 - 9 | 751 | 73 | ||
| U89488.1 | - | 9 - 10 | 999 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldb1
Genomic Map