Assay Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053100.2 | NP_444330.2 | 4 - 5 | 1732 | 96 | |
| GenBank mRNA | AF220035.1 | - | 3 - 4 | 351 | 96 | |
| AF281047.1 | - | 4 - 5 | 1081 | 96 | ||
| AK052763.1 | - | 4 - 5 | 467 | 96 | ||
| AK079847.1 | - | 2 - 3 | 171 | 96 | ||
| AK155303.1 | - | 4 - 5 | 1679 | 96 | ||
| BC029283.1 | - | 4 - 5 | 1666 | 96 | ||
| BC037065.1 | - | 4 - 5 | 1017 | 96 | ||
| BC060094.1 | - | 4 - 5 | 962 | 96 | ||
| BC082298.1 | - | 4 - 5 | 1607 | 96 | ||
| BC152343.1 | - | 4 - 5 | 1068 | 40093266 | 96 | |
| BC152344.1 | - | 4 - 5 | 1068 | 40093269 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim8
Genomic Map