Assay Details
| Gene Symbol: Elovl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007703.2 | NP_031729.1 | 1 - 2 | 275 | 78 | |
| GenBank mRNA | BC016468.1 | - | 1 - 2 | 270 | 4196583 | 78 |
| U97107.1 | - | 1 - 2 | 275 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elovl3
Genomic Map