Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029186.2 | NP_083462.2 | 5 - 6 | 1009 | 86 | |
| GenBank mRNA | AK015988.1 | - | 2 - 3 | 366 | 86 | |
| AK031554.1 | - | 5 - 6 | 955 | 86 | ||
| AK052797.1 | - | 2 - 3 | 167 | 86 | ||
| AK151797.1 | - | 5 - 6 | 984 | 86 | ||
| AK161305.1 | - | 5 - 6 | 965 | 86 | ||
| AK170247.1 | - | 956 | 86 | |||
| BC026385.1 | - | 3 - 4 | 1137 | 86 | ||
| BC031169.1 | - | 5 - 6 | 954 | 86 | ||
| BC049142.1 | - | 2 - 3 | 295 | 86 | ||
| BC058746.1 | - | 5 - 6 | 955 | 6399588 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem180
Genomic Map