Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029186.2 | NP_083462.2 | 7 - 8 | 1281 | 60 | |
| GenBank mRNA | AK015988.1 | - | 4 - 5 | 637 | 60 | |
| AK031554.1 | - | 7 - 8 | 1227 | 60 | ||
| AK151797.1 | - | 7 - 8 | 1256 | 60 | ||
| AK161305.1 | - | 7 - 8 | 1237 | 60 | ||
| AK170247.1 | - | 1228 | 60 | |||
| BC049142.1 | - | 4 - 5 | 567 | 60 | ||
| BC058746.1 | - | 7 - 8 | 1227 | 6399588 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem180
Genomic Map