Assay Details
| Gene Symbol: Psd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028627.2 | NP_082903.2 | 2 - 3 | 128 | 81 | |
| GenBank mRNA | BC057353.1 | - | 2 - 3 | 128 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_028627.2 transcripts.
Set Membership: |
|

Psd
Genomic Map