Assay Details
| Gene Symbol: Psd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028627.2 | NP_082903.2 | 4 - 5 | 974 | 82 | |
| GenBank mRNA | BC057353.1 | - | 4 - 5 | 974 | 82 | |
| BC138652.1 | - | 2 - 3 | 833 | 8861672 | 82 | |
| BC145352.1 | - | 2 - 3 | 833 | 9053908 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psd
Genomic Map