Assay Details
| Gene Symbol: Cdkn2aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172407.3 | NP_765995.1 | 1 - 2 | 410 | 85 | |
| GenBank mRNA | AK048437.1 | - | 1 - 2 | 379 | 85 | |
| AK078329.1 | - | 1 - 2 | 408 | 85 | ||
| AK089057.1 | - | 1 - 2 | 411 | 85 | ||
| AK133831.1 | - | 1 - 2 | 410 | 85 | ||
| AK167684.1 | - | 1 - 2 | 409 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdkn2aip
Genomic Map