Assay Details
| Gene Symbol: Adcy8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291903.1 | NP_001278832.1 | 13 - 14 | 4014 | 65 | |
| NM_009623.2 | NP_033753.2 | 14 - 15 | 4104 | 65 | ||
| GenBank mRNA | AK138244.1 | - | 13 - 14 | 4015 | 65 | |
| BC059010.1 | - | 14 - 15 | 3201 | 65 | ||
| BC094231.1 | - | 14 - 15 | 3542 | 65 | ||
| U85021.1 | - | 14 - 15 | 3964 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcy8
Genomic Map