Assay Details
| Gene Symbol: Phf20l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081409.1 | NP_001074878.1 | 5 - 6 | 745 | 62 | |
| GenBank mRNA | AK047044.1 | - | 5 - 6 | 758 | 62 | |
| BC082777.1 | - | 5 - 6 | 737 | 62 | ||
| BC145689.1 | - | 4 - 5 | 476 | 40130992 | 62 | |
More Information
Set Membership: |
|

Phf20l1
Genomic Map