Assay Details
| Gene Symbol: Phf20l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081409.1 | NP_001074878.1 | 6 - 7 | 822 | 107 | |
| GenBank mRNA | AK032638.1 | - | 6 - 7 | 837 | 107 | |
| AK047044.1 | - | 6 - 7 | 835 | 107 | ||
| BC145689.1 | - | 5 - 6 | 553 | 40130992 | 107 | |
More Information
Set Membership: |
|

Phf20l1
Genomic Map