Assay Details
| Gene Symbol: Phf20l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081409.1 | NP_001074878.1 | 9 - 10 | 1240 | 100 | |
| GenBank mRNA | AK032638.1 | - | 9 - 10 | 1255 | 100 | |
| BC061492.1 | - | 8 - 9 | 1106 | 100 | ||
| BC145689.1 | - | 8 - 9 | 971 | 40130992 | 100 | |
More Information
Set Membership: |
|

Phf20l1
Genomic Map