Assay Details
| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023232.3 | NP_075721.3 | 3 - 4 | 345 | 124 | |
| GenBank mRNA | AF203914.1 | - | 3 - 4 | 311 | 124 | |
| AK002887.1 | - | 3 - 4 | 315 | 124 | ||
| AK012760.1 | - | 3 - 4 | 343 | 124 | ||
| AK077198.1 | - | 3 - 4 | 326 | 124 | ||
| AK082903.1 | - | 3 - 4 | 315 | 124 | ||
| AK133194.1 | - | 325 | 124 | |||
| AK169573.1 | - | 3 - 4 | 315 | 124 | ||
| BC024780.1 | - | 3 - 4 | 317 | 5365266 | 124 | |
| BC054109.1 | - | 3 - 4 | 339 | 30013833 | 124 | |
More Information
Set Membership: |
|

Diablo
Genomic Map