Assay Details
| Gene Symbol: Hpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK149416.1 | - | 1 - 2 | 50 | 80 | |
| D29987.1 | - | 1 - 2 | 50 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149416.1, D29987.1 transcripts.
Set Membership: |
|

Hpd
Genomic Map