Assay Details
| Gene Symbol: Ext1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010162.2 | NP_034292.2 | 5 - 6 | 2235 | 94 | |
| GenBank mRNA | AK132327.1 | - | 5 - 6 | 2212 | 94 | |
| BC004741.1 | - | 5 - 6 | 1599 | 3499743 | 94 | |
| U78539.1 | - | 5 - 6 | 1739 | 94 | ||
| X96639.1 | - | 5 - 6 | 2009 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ext1
Genomic Map