Assay Details
| Gene Symbol: Wwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133791.4 | NP_598552.2 | 13 - 14 | 2463 | 109 | |
| GenBank mRNA | AB045323.1 | - | 13 - 14 | 2488 | 109 | |
| AK028956.1 | - | 80 | 109 | |||
| AK032989.1 | - | 9 - 10 | 1529 | 109 | ||
| AK079375.1 | - | 13 - 14 | 2299 | 109 | ||
| AK147648.1 | - | 2457 | 109 | |||
| BC054434.1 | - | 3 - 4 | 744 | 109 | ||
| BC067050.1 | - | 13 - 14 | 2463 | 30537299 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wwc2
Genomic Map