Assay Details
| Gene Symbol: Wwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133791.4 | NP_598552.2 | 14 - 15 | 2551 | 76 | |
| GenBank mRNA | AB045323.1 | - | 14 - 15 | 2576 | 76 | |
| AK028956.1 | - | 168 | 76 | |||
| AK032989.1 | - | 10 - 11 | 1617 | 76 | ||
| AK079375.1 | - | 14 - 15 | 2387 | 76 | ||
| AK147648.1 | - | 2545 | 76 | |||
| BC054434.1 | - | 4 - 5 | 832 | 76 | ||
| BC067050.1 | - | 14 - 15 | 2551 | 30537299 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wwc2
Genomic Map