Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500012026.mm.1 |
NM_153548.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012026.mm.1 |
NM_153548.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012026.mm.1 |
TC1500001596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012025.mm.1 |
NM_153548.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012025.mm.1 |
NM_153548.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012025.mm.1 |
TC1500001596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012027.mm.1 |
NM_153548.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012027.mm.1 |
NM_153548.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012027.mm.1 |
TC1500001596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012023.mm.1 |
NM_153548.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012023.mm.1 |
NM_153548.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012023.mm.1 |
TC1500001596.mm.1 |
MTA TC |