Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 20 - 21 | 2737 | 66 | |
| GenBank mRNA | AK088754.1 | - | 5 - 6 | 610 | 66 | |
| AK128970.1 | - | 20 - 21 | 2752 | 66 | ||
| AK163968.1 | - | 3 - 4 | 361 | 66 | ||
| BC031364.1 | - | 6 - 7 | 751 | 66 | ||
| BC034070.1 | - | 10 - 11 | 1337 | 4217923 | 66 | |
| BC040815.1 | - | 10 - 11 | 1337 | 4217923 | 66 | |
| BC067035.1 | - | 20 - 21 | 2737 | 30532658 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map