Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 25 - 26 | 3316 | 61 | |
| GenBank mRNA | AK088754.1 | - | 10 - 11 | 1189 | 61 | |
| AK128970.1 | - | 25 - 26 | 3331 | 61 | ||
| AK129086.1 | - | 2968 | 61 | |||
| AK163968.1 | - | 8 - 9 | 940 | 61 | ||
| BC031364.1 | - | 11 - 12 | 1330 | 61 | ||
| BC034070.1 | - | 15 - 16 | 1916 | 4217923 | 61 | |
| BC040815.1 | - | 15 - 16 | 1916 | 4217923 | 61 | |
| BC067035.1 | - | 25 - 26 | 3316 | 30532658 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map