Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 2 - 3 | 82 | ||
| GenBank mRNA | AK128970.1 | - | 2 - 3 | 82 | ||
| AK215294.1 | - | 1 - 2 | 82 | |||
| BC067035.1 | - | 2 - 3 | 30532658 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map