Assay Details
| Gene Symbol: Mtss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146180.1 | NP_001139652.1 | 3 - 4 | 709 | 57 | |
| NM_144800.2 | NP_659049.2 | 3 - 4 | 709 | 57 | ||
| GenBank mRNA | AK129139.1 | - | 3 - 4 | 706 | 57 | |
| AY214918.1 | - | 3 - 4 | 210 | 57 | ||
| BC024131.1 | - | 3 - 4 | 674 | 5101888 | 57 | |
More Information
Set Membership: |
|

Mtss1
Genomic Map