Assay Details
| Gene Symbol: Nmrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145497.2 | NP_663472.1 | 3 - 4 | 224 | 93 | |
| GenBank mRNA | AK050579.1 | - | 3 - 4 | 255 | 93 | |
| AK169962.1 | - | 3 - 4 | 198 | 93 | ||
| BC016495.1 | - | 3 - 4 | 224 | 4221437 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmrk1
Genomic Map