Assay Details
| Gene Symbol: Irf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008391.4 | NP_032417.3 | 6 - 7 | 733 | 114 | |
| GenBank mRNA | AK036652.1 | - | 6 - 7 | 701 | 114 | |
| AK132099.1 | - | 6 - 7 | 746 | 114 | ||
| AK155025.1 | - | 6 - 7 | 718 | 114 | ||
| BC006577.1 | - | 6 - 7 | 726 | 3586586 | 114 | |
| BC110666.1 | - | 6 - 7 | 705 | 4918447 | 114 | |
| J03168.1 | - | 6 - 7 | 688 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf2
Genomic Map