| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800016713.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016713.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016713.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016713.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016714.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016714.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016714.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016714.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016711.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016711.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016711.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016711.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016711.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016711.mm.1 |
TC0800002169.mm.1 |
MTA TC |