| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800016698.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016698.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016698.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016698.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016698.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016698.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016699.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016699.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016699.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016699.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016699.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016699.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016706.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016706.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016706.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016706.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016706.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016706.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016705.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016705.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016705.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016705.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016705.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016705.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016702.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016702.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016702.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016702.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016702.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016702.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016695.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016695.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016695.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016695.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016695.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016695.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016701.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016701.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016701.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016701.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016701.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016701.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016703.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016703.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016703.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016703.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016703.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016703.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016704.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016704.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016704.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016704.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016704.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016704.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016696.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016696.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016696.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016696.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016696.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016696.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016693.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016693.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016693.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016693.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016693.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016693.mm.1 |
TC0800002169.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016700.mm.1 |
NM_001114311.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016700.mm.1 |
NM_001114311.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016700.mm.1 |
NM_001286376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016700.mm.1 |
NM_175162.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016700.mm.1 |
NM_175162.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016700.mm.1 |
TC0800002169.mm.1 |
MTA TC |