| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500014105.mm.1 |
NM_001081970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014105.mm.1 |
NM_001081970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014105.mm.1 |
TC1500001818.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014108.mm.1 |
NM_001081970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014108.mm.1 |
NM_001081970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014108.mm.1 |
TC1500001818.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014104.mm.1 |
NM_001081970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014104.mm.1 |
NM_001081970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014104.mm.1 |
TC1500001818.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014107.mm.1 |
NM_001081970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014107.mm.1 |
NM_001081970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014107.mm.1 |
TC1500001818.mm.1 |
MTA TC |