Assay Details
| Gene Symbol: Deptor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037937.3 | NP_001033026.1 | 2 - 3 | 300 | 82 | |
| NM_145470.3 | NP_663445.2 | 2 - 3 | 478 | 82 | ||
| GenBank mRNA | AK139888.1 | - | 2 - 3 | 300 | 82 | |
| AK145208.1 | - | 2 - 3 | 372 | 82 | ||
| AK220300.1 | - | 2 - 3 | 312 | 82 | ||
| BC004774.1 | - | 2 - 3 | 483 | 82 | ||
| DV649395.1 | - | 2 - 3 | 445 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Deptor
Genomic Map