Assay Details
| Gene Symbol: Deptor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037937.3 | NP_001033026.1 | 3 - 4 | 430 | 71 | |
| NM_145470.3 | NP_663445.2 | 3 - 4 | 608 | 71 | ||
| GenBank mRNA | AK139888.1 | - | 3 - 4 | 430 | 71 | |
| AK220300.1 | - | 3 - 4 | 442 | 71 | ||
| BC004774.1 | - | 3 - 4 | 613 | 71 | ||
| DV649395.1 | - | 3 - 4 | 575 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Deptor
Genomic Map