Assay Details
| Gene Symbol: Sntb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016667.3 | NP_057876.1 | 5 - 6 | 1691 | 56 | |
| GenBank mRNA | U89997.1 | - | 5 - 6 | 1696 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sntb1
Genomic Map