Assay Details
| Gene Symbol: Mrpl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026759.3 | NP_081035.1 | 3 - 4 | 267 | 142 | |
| GenBank mRNA | AB049641.1 | - | 3 - 4 | 245 | 142 | |
| AK003283.2 | - | 3 - 4 | 268 | 142 | ||
| BC055470.1 | - | 3 - 4 | 231 | 142 | ||
| BC060968.1 | - | 3 - 4 | 273 | 6741102 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl13
Genomic Map