Assay Details
| Gene Symbol: Mrpl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026759.3 | NP_081035.1 | 4 - 5 | 334 | 60 | |
| GenBank mRNA | AB049641.1 | - | 4 - 5 | 312 | 60 | |
| AK003283.2 | - | 4 - 5 | 335 | 60 | ||
| AK160744.1 | - | 3 - 4 | 237 | 60 | ||
| BC027542.1 | - | 3 - 4 | 224 | 60 | ||
| BC048498.1 | - | 3 - 4 | 243 | 60 | ||
| BC055470.1 | - | 4 - 5 | 298 | 60 | ||
| BC060968.1 | - | 4 - 5 | 340 | 6741102 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC027542.1 transcripts.
Set Membership: |
|

Mrpl13
Genomic Map