| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081462.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081462.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081463.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081463.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081464.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081464.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081465.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081465.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081466.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081466.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081467.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081467.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081468.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081468.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081469.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081469.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081470.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001081470.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001244936.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_001244936.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_020331.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
NM_020331.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500030396.mm.1 |
TC0500003371.mm.1 |
MTA TC |