Assay Details
| Gene Symbol: Tbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011535.3 | NP_035665.2 | 2 - 3 | 1444 | 95 | |
| GenBank mRNA | AK052453.1 | - | 2 - 3 | 1659 | 95 | |
| AK145436.1 | - | 2 - 3 | 1361 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbx3
Genomic Map