Assay Details
| Gene Symbol: Tbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011535.3 | NP_035665.2 | 3 - 4 | 1496 | 80 | |
| GenBank mRNA | AK052453.1 | - | 3 - 4 | 1711 | 80 | |
| AK145436.1 | - | 3 - 4 | 1413 | 80 | ||
More Information
Set Membership: |
|

Tbx3
Genomic Map