Assay Details
| Gene Symbol: Tbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011535.3 | NP_035665.2 | 7 - 8 | 2551 | 65 | |
| NM_198052.2 | NP_932169.1 | 7 - 8 | 2687 | 65 | ||
| GenBank mRNA | AK052453.1 | - | 7 - 8 | 2766 | 65 | |
| AK087128.1 | - | 1 - 2 | 319 | 65 | ||
| AK145436.1 | - | 7 - 8 | 2468 | 65 | ||
| AK157247.1 | - | 6 - 7 | 2252 | 65 | ||
| BC096551.1 | - | 7 - 8 | 2687 | 30547736 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbx3
Genomic Map