Assay Details
| Gene Symbol: Il33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164724.1 | NP_001158196.1 | 3 - 4 | 310 | 106 | |
| NM_133775.2 | NP_598536.2 | 3 - 4 | 289 | 106 | ||
| GenBank mRNA | AK020353.1 | - | 3 - 4 | 290 | 106 | |
| AK075849.1 | - | 3 - 4 | 311 | 106 | ||
| AK163464.1 | - | 3 - 4 | 290 | 106 | ||
| AY905582.1 | - | 2 - 3 | 230 | 106 | ||
| BC003847.1 | - | 3 - 4 | 266 | 3593927 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il33
Genomic Map