Assay Details
| Gene Symbol: Slc1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009199.2 | NP_033225.1 | 10 - 11 | 1262 | 72 | |
| GenBank mRNA | AK018188.1 | - | 10 - 11 | 1295 | 72 | |
| AK083097.1 | - | 10 - 11 | 1297 | 72 | ||
| BC065099.1 | - | 10 - 11 | 1286 | 30364069 | 72 | |
| D43797.1 | - | 10 - 11 | 1268 | 72 | ||
| U73521.1 | - | 10 - 11 | 1262 | 72 | ||
| U75214.1 | - | 6 - 7 | 1037 | 72 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AF087578.1 | - | Chr. - - [-] on Build | 72 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a1
Genomic Map