Assay Details
| Gene Symbol: Crybb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020891.1 | - | 1 - 2 | 29 | 60 | |
| BC085185.1 | - | 2 - 3 | 171 | 30679384 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK020891.1, BC085185.1 transcripts.
Set Membership: |
|

Crybb2
Genomic Map