Assay Details
| Gene Symbol: Ide | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031156.3 | NP_112419.3 | 17 - 18 | 2234 | 86 | |
| GenBank mRNA | AJ278422.1 | - | 17 - 18 | 2254 | 86 | |
| AK004972.2 | - | 17 - 18 | 2236 | 86 | ||
| AK128986.1 | - | 17 - 18 | 2251 | 86 | ||
| BC041675.1 | - | 17 - 18 | 2266 | 4951971 | 86 | |
More Information
Set Membership: |
|

Ide
Genomic Map